Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pfkma | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004575.2 | NP_001004575.1 | 16 - 17 | 1744 | 70 | |
GenBank mRNA | BC081652.1 | - | 16 - 17 | 1720 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |