Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: osbpl3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004646.1 | NP_001004646.1 | 10 - 11 | 1044 | 66 | |
XM_005170028.4 | XP_005170085.1 | 10 - 11 | 1298 | 66 | ||
GenBank mRNA | BC078429.1 | - | 10 - 11 | 1044 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |