Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gpsm2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005936.1 | NP_001005936.1 | 10 - 11 | 1375 | 81 | |
XM_021469788.1 | XP_021325463.1 | 10 - 11 | 81 | |||
GenBank mRNA | BC083520.1 | - | 10 - 11 | 1375 | 81 | |
DQ235092.1 | - | 9 - 10 | 1222 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |