Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: st6galnac5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006662.2 | NP_001006663.2 | 3 - 4 | 741 | 82 | |
XM_021478236.1 | XP_021333911.1 | 3 - 4 | 688 | 82 | ||
GenBank mRNA | AJ705083.1 | - | 1 - 2 | 422 | 82 | |
BC081635.1 | - | 3 - 4 | 718 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |