Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hmgcrb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014292.2 | NP_001014314.1 | 18 - 19 | 2568 | 63 | |
GenBank mRNA | BC091792.1 | - | 18 - 19 | 2572 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |