Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc5a8l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034969.1 | NP_001030141.1 | 3 - 4 | 577 | 66 | |
GenBank mRNA | AY727859.1 | - | 3 - 4 | 577 | 66 | |
BC124585.1 | - | 3 - 4 | 484 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |