Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: stc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045457.1 | NP_001038922.1 | 2 - 3 | 411 | 75 | |
GenBank mRNA | BC122228.1 | - | 2 - 3 | 411 | 75 | |
BC165445.1 | - | 2 - 3 | 288 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |