Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: erbin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193543.1 | NP_001180472.1 | 5 - 6 | 927 | 78 | |
XM_005155576.4 | XP_005155633.1 | 5 - 6 | 1185 | 78 | ||
XM_005155577.3 | XP_005155634.1 | 5 - 6 | 927 | 78 | ||
XM_005155578.4 | XP_005155635.1 | 5 - 6 | 1181 | 78 | ||
GenBank mRNA | BC125845.1 | - | 4 - 5 | 840 | 78 | |
BC146713.1 | - | 5 - 6 | 927 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |