Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
TaqMan
®
Gene Expression Assays
›
Search Tool
›
Search Results
› Dr03130638_m1
See other PTPRFB GE Assays ›
Gene Symbol:
ptprfb
Gene Name:
protein tyrosine phosphatase receptor type Fb
Gene Aliases:
fj23e12, lar, ptprf, si:dkey-21k10.1, wu:fj23e12
Chromosome Location:
Chr.2: 17578415 - 17947246 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID
Dr03130638_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: ptprfb
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001083576.1
NP_001077045.1
2 - 3
465
67
XM_009298457.3
XP_009296732.1
2 - 3
513
67
XM_009298458.3
XP_009296733.1
2 - 3
301
67
XM_009298459.3
XP_009296734.1
2 - 3
513
67
XM_009298460.3
XP_009296735.1
2 - 3
513
67
XM_009298461.3
XP_009296736.1
2 - 3
513
67
XM_009298462.3
XP_009296737.1
2 - 3
513
67
XM_009298463.3
XP_009296738.1
2 - 3
513
67
XM_009298464.3
XP_009296739.1
2 - 3
513
67
XM_009298466.3
XP_009296741.1
2 - 3
512
67
XM_009298467.3
XP_009296742.1
2 - 3
512
67
XM_009298468.3
XP_009296743.1
2 - 3
512
67
XM_009298469.3
XP_009296744.1
2 - 3
512
67
XM_021481410.1
XP_021337085.1
2 - 3
838
67
XM_021481419.1
XP_021337094.1
2 - 3
513
67
XM_021481437.1
XP_021337112.1
2 - 3
512
67
XM_021481438.1
XP_021337113.1
2 - 3
512
67
XM_021481440.1
XP_021337115.1
2 - 3
512
67
XM_021481441.1
XP_021337116.1
2 - 3
512
67
XM_021481443.1
XP_021337118.1
2 - 3
512
67
GenBank mRNA
BC134804.1
-
2 - 3
465
67
Back To Top
More Information
Set Membership:
Amplicon length less than or equal to 70
Mammalian Gene Collection
Danio rerio
Probe spans exons
Gene Ontology Categories:
Function(s)
Process(es)
protein dephosphorylation
cell adhesion
dephosphorylation
peptidyl-tyrosine dephosphorylation
sensory neuron axon guidance
synaptic membrane adhesion
phosphoprotein phosphatase activity
protein tyrosine phosphatase activity
heparin binding
hydrolase activity
phosphatase activity
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-x5whk:80/10.239.163.84:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0