Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131227.1 | NP_571302.1 | 1 - 2 | 240 | 94 | |
GenBank mRNA | AF001909.1 | - | 1 - 2 | 240 | 94 | |
BC163788.1 | - | 1 - 2 | 326 | 94 | ||
BC163791.1 | - | 1 - 2 | 326 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Danio rerio Inventoried Probe spans exons |