Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gfra1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131731.1 | NP_571806.1 | 5 - 6 | 914 | 67 | |
GenBank mRNA | AY436321.1 | - | 5 - 6 | 914 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Danio rerio Probe spans exons |