Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cry4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131787.1 | NP_571862.1 | 8 - 9 | 1380 | 59 | |
XM_005168277.4 | XP_005168334.1 | 9 - 10 | 1607 | 59 | ||
XM_009295766.3 | XP_009294041.1 | 9 - 10 | 1594 | 59 | ||
GenBank mRNA | AB042253.1 | - | 8 - 9 | 1380 | 59 | |
BC044385.1 | - | 9 - 10 | 1514 | 59 | ||
BC097079.1 | - | 7 - 8 | 1124 | 59 | ||
BC164413.1 | - | 9 - 10 | 1477 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |