Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rbpja | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198878.2 | NP_942579.2 | 10 - 11 | 1181 | 92 | |
GenBank mRNA | AY128532.1 | - | 10 - 11 | 1187 | 92 | |
BC163922.1 | - | 10 - 11 | 1181 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |