Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rbpja | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198878.2 | NP_942579.2 | 4 - 5 | 346 | 92 | |
GenBank mRNA | AY128532.1 | - | 4 - 5 | 346 | 92 | |
AY332617.1 | - | 4 - 5 | 319 | 92 | ||
BC092918.1 | - | 4 - 5 | 323 | 92 | ||
BC163922.1 | - | 4 - 5 | 346 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |