Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rps6kal | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200073.1 | NP_956367.1 | 2 - 3 | 238 | 87 | |
GenBank mRNA | BC045478.1 | - | 2 - 3 | 342 | 87 | |
BC057467.1 | - | 2 - 3 | 238 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |