Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: wrnip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212920.1 | NP_998085.1 | 2 - 3 | 779 | 118 | |
GenBank mRNA | BC067729.1 | - | 2 - 3 | 779 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |