Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gnb3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213202.1 | NP_998367.1 | 7 - 8 | 876 | 82 | |
XM_005159345.4 | XP_005159402.1 | 7 - 8 | 885 | 82 | ||
GenBank mRNA | BC059436.1 | - | 7 - 8 | 82 | ||
BC065866.1 | - | 7 - 8 | 876 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Inventoried Probe spans exons |