Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: asmtl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213511.1 | NP_998676.1 | 10 - 11 | 1301 | 89 | |
GenBank mRNA | BC055218.1 | - | 10 - 11 | 1301 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |