Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ssr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002082.1 | NP_001002082.1 | 4 - 5 | 403 | 144 | |
GenBank mRNA | BC071553.1 | - | 4 - 5 | 403 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |