Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ctdnep1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007440.2 | NP_001007441.2 | 2 - 3 | 656 | 138 | |
GenBank mRNA | BC058872.1 | - | 2 - 3 | 452 | 138 | |
BC085403.1 | - | 2 - 3 | 452 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |