Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cd2ap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008583.2 | NP_001008583.2 | 3 - 4 | 624 | 77 | |
XM_005160684.4 | XP_005160741.1 | 2 - 3 | 255 | 77 | ||
XM_009294709.3 | XP_009292984.1 | 3 - 4 | 655 | 77 | ||
GenBank mRNA | BC086846.1 | - | 2 - 3 | 483 | 77 | |
BC164631.1 | - | 2 - 3 | 294 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |