Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pih1d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008629.1 | NP_001008629.1 | 2 - 3 | 544 | 63 | |
XM_009291562.1 | XP_009289837.1 | 2 - 3 | 542 | 63 | ||
XM_021481399.1 | XP_021337074.1 | 2 - 3 | 539 | 63 | ||
XM_021481401.1 | XP_021337076.1 | 2 - 3 | 539 | 63 | ||
XM_021481402.1 | XP_021337077.1 | 2 - 3 | 539 | 63 | ||
XM_021481403.1 | XP_021337078.1 | 2 - 3 | 140 | 63 | ||
GenBank mRNA | BC086728.1 | - | 2 - 3 | 544 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |