Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rps24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012316.1 | NP_001012316.1 | 4 - 5 | 427 | 112 | |
XM_021480547.1 | XP_021336222.1 | 4 - 5 | 633 | 112 | ||
GenBank mRNA | BC081494.1 | - | 4 - 5 | 427 | 112 | |
BC094973.1 | - | 4 - 5 | 427 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |