Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gli2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015069.1 | NP_001015069.1 | 7 - 8 | 1309 | 57 | |
GenBank mRNA | AY928397.2 | - | 7 - 8 | 1309 | 57 | |
BC162463.1 | - | 8 - 9 | 1106 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |