Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gm2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017831.1 | NP_001017831.1 | 3 - 4 | 402 | 73 | |
XM_005173119.4 | XP_005173176.1 | 3 - 4 | 439 | 73 | ||
GenBank mRNA | BC092784.1 | - | 3 - 4 | 402 | 73 | |
BC165264.1 | - | 2 - 3 | 293 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |