Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cuedc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017994.2 | NP_001017994.1 | 4 - 5 | 529 | 83 | |
XM_005156782.2 | XP_005156839.1 | 5 - 6 | 601 | 83 | ||
GenBank mRNA | AY394932.1 | - | 5 - 6 | 610 | 83 | |
BC081633.1 | - | 4 - 5 | 529 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |