Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: styx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024390.1 | NP_001019561.1 | 7 - 8 | 546 | 89 | |
GenBank mRNA | BC095339.1 | - | 7 - 8 | 546 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |