Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sinhcaf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025531.1 | NP_001020702.1 | 5 - 6 | 535 | 73 | |
GenBank mRNA | BC162866.1 | - | 5 - 6 | 535 | 73 | |
BC162884.1 | - | 5 - 6 | 535 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |