Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: smtlb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037674.1 | NP_001032763.1 | 4 - 5 | 536 | 102 | |
GenBank mRNA | AJ867249.1 | - | 4 - 5 | 533 | 102 | |
AY221126.1 | - | 4 - 5 | 536 | 102 | ||
BC121735.1 | - | 4 - 5 | 538 | 102 | ||
BC165060.1 | - | 4 - 5 | 535 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |