Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: asap2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045207.1 | NP_001038672.1 | 16 - 17 | 1974 | 81 | |
XM_005158734.4 | XP_005158791.1 | 17 - 18 | 2004 | 81 | ||
XM_021467217.1 | XP_021322892.1 | 18 - 19 | 2031 | 81 | ||
XM_021467218.1 | XP_021322893.1 | 17 - 18 | 2022 | 81 | ||
XM_021467219.1 | XP_021322894.1 | 18 - 19 | 2031 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Danio rerio Probe spans exons |