Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: il22ra2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045279.1 | NP_001038744.1 | 1 - 2 | 79 | 92 | |
GenBank mRNA | BC116595.1 | - | 1 - 2 | 79 | 92 | |
EF014959.1 | - | 1 - 2 | 100 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Danio rerio Inventoried Probe spans exons |