Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: usp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077449.1 | NP_001070917.1 | 29 - 30 | 4590 | 78 | |
XM_005167736.4 | XP_005167793.1 | 30 - 31 | 5084 | 78 | ||
XM_009304755.3 | XP_009303030.1 | 30 - 31 | 5085 | 78 | ||
XM_009304756.3 | XP_009303031.1 | 30 - 31 | 5085 | 78 | ||
XM_009304757.3 | XP_009303032.1 | 30 - 31 | 5086 | 78 | ||
GenBank mRNA | DQ086492.1 | - | 29 - 30 | 4590 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Danio rerio Probe spans exons |