Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nt5c2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079966.2 | NP_001073435.2 | 10 - 11 | 968 | 92 | |
XM_005168392.4 | XP_005168449.1 | 10 - 11 | 1137 | 92 | ||
XM_021474992.1 | XP_021330667.1 | 9 - 10 | 812 | 92 | ||
XM_021474993.1 | XP_021330668.1 | 5 - 6 | 330 | 92 | ||
GenBank mRNA | BC076085.1 | - | 10 - 11 | 1067 | 92 | |
BC124713.1 | - | 10 - 11 | 913 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |