Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nt5c2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079966.2 | NP_001073435.2 | 15 - 16 | 1407 | 71 | |
XM_005168392.4 | XP_005168449.1 | 15 - 16 | 1576 | 71 | ||
XM_021474992.1 | XP_021330667.1 | 14 - 15 | 1251 | 71 | ||
XM_021474993.1 | XP_021330668.1 | 10 - 11 | 769 | 71 | ||
GenBank mRNA | BC076085.1 | - | 15 - 16 | 1506 | 71 | |
BC124713.1 | - | 15 - 16 | 1352 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |