Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mfge8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080990.1 | NP_001074459.1 | 2 - 3 | 268 | 75 | |
XM_005163016.4 | XP_005163073.1 | 2 - 3 | 333 | 75 | ||
GenBank mRNA | BC131837.1 | - | 2 - 3 | 268 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |