Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sptan1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098488.1 | NP_001091958.1 | 4 - 5 | 588 | 64 | |
XM_021468823.1 | XP_021324498.1 | 3 - 4 | 508 | 64 | ||
XM_021468824.1 | XP_021324499.1 | 4 - 5 | 640 | 64 | ||
XM_021468825.1 | XP_021324500.1 | 4 - 5 | 719 | 64 | ||
XM_021468826.1 | XP_021324501.1 | 4 - 5 | 526 | 64 | ||
XM_021468827.1 | XP_021324502.1 | 3 - 4 | 508 | 64 | ||
XM_021468828.1 | XP_021324503.1 | 3 - 4 | 508 | 64 | ||
XM_021468829.1 | XP_021324504.1 | 3 - 4 | 508 | 64 | ||
XM_021468830.1 | XP_021324505.1 | 3 - 4 | 508 | 64 | ||
XM_021468831.1 | XP_021324506.1 | 3 - 4 | 508 | 64 | ||
XM_021468832.1 | XP_021324507.1 | 3 - 4 | 508 | 64 | ||
XM_021468833.1 | XP_021324508.1 | 3 - 4 | 508 | 64 | ||
XM_021468834.1 | XP_021324509.1 | 3 - 4 | 508 | 64 | ||
GenBank mRNA | EF375552.1 | - | 4 - 5 | 588 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Danio rerio Probe spans exons |