Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: poglut3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098745.1 | NP_001092215.1 | 2 - 3 | 645 | 83 | |
GenBank mRNA | BC142839.1 | - | 2 - 3 | 645 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |