Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kcng4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110029.1 | NP_001103499.1 | 2 - 3 | 998 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Danio rerio Probe spans exons |