Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hoxa9a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131532.2 | NP_571607.2 | 1 - 2 | 593 | 125 | |
GenBank mRNA | BC090700.1 | - | 1 - 2 | 593 | 125 | |
BC165442.1 | - | 1 - 2 | 546 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Probe spans exons |