Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tdo2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199856.1 | NP_956150.1 | 5 - 6 | 502 | 82 | |
XM_021479554.1 | XP_021335229.1 | 5 - 6 | 506 | 82 | ||
GenBank mRNA | BC055123.1 | - | 5 - 6 | 502 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |