Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: skp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200743.1 | NP_957037.1 | 4 - 5 | 359 | 76 | |
GenBank mRNA | AY648843.1 | - | 3 - 4 | 339 | 76 | |
BC059536.1 | - | 4 - 5 | 359 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |