Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nrp2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212965.1 | NP_998130.1 | 7 - 8 | 1761 | 72 | |
XM_021469092.1 | XP_021324767.1 | 7 - 8 | 1781 | 72 | ||
GenBank mRNA | AY351912.1 | - | 7 - 8 | 1497 | 72 | |
AY428801.1 | - | 7 - 8 | 1761 | 72 | ||
AY437405.1 | - | 7 - 8 | 1747 | 72 | ||
BC162118.1 | - | 7 - 8 | 1643 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |