Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cox4i1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214701.1 | NP_999866.1 | 4 - 5 | 408 | 95 | |
GenBank mRNA | AY391417.1 | - | 4 - 5 | 408 | 95 | |
BC095163.1 | - | 4 - 5 | 415 | 95 | ||
BC164879.1 | - | 3 - 4 | 408 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |