Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: psmd4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002112.2 | NP_001002112.1 | 7 - 8 | 775 | 91 | |
GenBank mRNA | BC071482.1 | - | 7 - 8 | 776 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |