Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: psmc3ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002124.2 | NP_001002124.1 | 3 - 4 | 301 | 75 | |
GenBank mRNA | BC071453.1 | - | 3 - 4 | 303 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |