Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sav1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004560.1 | NP_001004560.1 | 3 - 4 | 1153 | 107 | |
GenBank mRNA | BC081632.1 | - | 3 - 4 | 1153 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |