Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: reep6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004656.2 | NP_001004656.1 | 3 - 4 | 443 | 99 | |
XM_009305809.3 | XP_009304084.1 | 3 - 4 | 477 | 99 | ||
XM_021479599.1 | XP_021335274.1 | 3 - 4 | 477 | 99 | ||
GenBank mRNA | BC081377.1 | - | 3 - 4 | 420 | 99 | |
BC165140.1 | - | 3 - 4 | 399 | 99 | ||
EH432211.1 | - | 3 - 4 | 475 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |