Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hgfa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013274.1 | NP_001013292.1 | 13 - 14 | 1682 | 121 | |
XM_005164742.3 | XP_005164799.1 | 14 - 15 | 2641 | 121 | ||
GenBank mRNA | BC135005.1 | - | 14 - 15 | 1718 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |