Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: wdr27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_009307487.3 | XP_009305762.1 | 4 - 5 | 532 | 63 | |
GenBank mRNA | BC109429.1 | - | 4 - 5 | 537 | 63 | |
DN599258.1 | - | 4 - 5 | 529 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |