Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: jph1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044348.1 | NP_001037813.1 | 3 - 4 | 1518 | 74 | |
XM_005163433.4 | XP_005163490.1 | 3 - 4 | 1546 | 74 | ||
GenBank mRNA | BC117663.1 | - | 3 - 4 | 1518 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |